Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-1-hepten-4-ol, 98%

Chemical Identifiers

Chemical Name or Material 4-Methyl-1-hepten-4-ol
CAS 1186-31-8
Molecular Formula C8H16O
Formula Weight 128.22
MDL Number MFCD00021838
UN Number UN1987
Synonym Allyl methyl n-propyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2082614

  SDS
alfa aesar™
B2082614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2082622

  SDS
alfa aesar™
B2082622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2082606

  SDS
alfa aesar™
B2082606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 4-Methyl-1-hepten-4-ol
Flash Point 51°C (124°F)
Quantity 25g
CAS 1186-31-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
Assay 98%
Molecular Formula C8H16O
Formula Weight 128.22
MDL Number MFCD00021838
UN Number UN1987
Synonym Allyl methyl n-propyl carbinol
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.