missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-1-cyclohexene, TCI America™

Manufacturer:  TCI America M0201500ML

 View more versions of this product

Catalog No. M0201500ML



Specifications

Specifications

4-Methyl-1-cyclohexene
Yellow
591-47-9
MFCD00001573
FSWCCQWDVGZMRD-UHFFFAOYSA-N
4-methylcyclohexene
11572
Clear Liquid at 20°C
104°C
500mL
C7H12
3295
CC1CCC=CC1
96.173
96.17

Chemical Identifiers

591-47-9
96.173
FSWCCQWDVGZMRD-UHFFFAOYSA-N
4-methylcyclohexene
C7H12
MFCD00001573
11572
CC1CCC=CC1
Safety and Handling

Safety and Handling

TSCA : Yes

Documents