Please login to your online account to display your discounted pricing

4-Methyl-1,3-thiazole-2-carboxylic acid, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC429360010

 View more versions of this product

Catalog No. AC429360010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 143.17
Quantity 1g
Color White
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
CAS 14542-16-6
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C5H5NO2S
Assay Percent Range 97%
MDL Number MFCD07377486
Chemical Name or Material 4-Methyl-1, 3-thiazole-2-carboxylic acid
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material 4-Methyl-1, 3-thiazole-2-carboxylic acid
CAS 14542-16-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H5NO2S
Formula Weight 143.17
MDL Number MFCD07377486
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents