missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-1,3-thiazole-2-carboxylic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC429360050

 View more versions of this product

Catalog No. AC429360050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
143.17
White
100.0
MFCD07377486
97%
C5H5NO2S
Crystalline Powder
5g
96.0
Glass bottle
4-Methyl-1, 3-thiazole-2-carboxylic acid
14542-16-6
Authentic

Chemical Identifiers

14542-16-6
100.0
C5H5NO2S
MFCD07377486
≥96.0%
96.0
97%
143.17
4-Methyl-1, 3-thiazole-2-carboxylic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.