missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methyl-1,3-oxazole-5-carboxylic acid, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC10001DA
Specifications
4-Methyl-1, 3-oxazole-5-carboxylic acid | |
95% | |
C5H5NO3 | |
4-methyloxazole-5-carboxylic acid, 4-methyl-5-oxazolecarboxylic acid, 5-oxazolecarboxylic acid, 4-methyl, 4-methyl-oxazole-5-carboxylic acid, ksc205g3h, 4-methyl-5-oxazolecarboxylicacid, 4-methyl-oxazol-5-carboxylic acid, 4-methoxyoxazole-5-carboxylic acid, 5-oxazolecarboxylicacid, 4-methyl | |
CC1=C(OC=N1)C(=O)O | |
127.099 | |
127.1 |
2510-32-9 | |
Amber Glass Bottle | |
1 g | |
ZIXUNDOOBLSXPE-UHFFFAOYSA-N | |
4-methyl-1,3-oxazole-5-carboxylic acid | |
292311 | |
95% |
Chemical Identifiers
2510-32-9 | |
127.099 | |
4-methyloxazole-5-carboxylic acid, 4-methyl-5-oxazolecarboxylic acid, 5-oxazolecarboxylic acid, 4-methyl, 4-methyl-oxazole-5-carboxylic acid, ksc205g3h, 4-methyl-5-oxazolecarboxylicacid, 4-methyl-oxazol-5-carboxylic acid, 4-methoxyoxazole-5-carboxylic acid, 5-oxazolecarboxylicacid, 4-methyl | |
4-methyl-1,3-oxazole-5-carboxylic acid |
C5H5NO3 | |
ZIXUNDOOBLSXPE-UHFFFAOYSA-N | |
292311 | |
CC1=C(OC=N1)C(=O)O |