missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-1,3-dioxane, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 305000500

 View more versions of this product

Catalog No. AC305000500


 

Specifications Description & Specifications

Specifications

4-Methyl-1, 3-dioxane
Clear colorless
99%
98.5
Authentic
1.4140 to 1.4160 (20°C, 589nm)
≥98.5%
C5H10O2
MFCD00006569
0.97g/mL
50g
1120-97-4
100.0
Glass bottle
Liquid
99%
102.13

Chemical Identifiers

1120-97-4
100.0
C5H10O2
MFCD00006569
≥98.5%
98.5
99%
102.13
4-Methyl-1, 3-dioxane, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 214-323-1

RTECSNumber : JH2200000