Please login to your online account to display your discounted pricing

4-Methyl-1,3-dioxane, 99%, ACROS Organics™

$98.21 - $98.21

Chemical Identifiers

Chemical Name or Material 4-Methyl-1, 3-dioxane
Name Note 99%
CAS 1120-97-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C5H10O2
Formula Weight 102.13
MDL Number MFCD00006569
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC305000500

  SDS
thermo fisher scientific
305000500
Glass bottle 50g Each for $98.21

AC305000050

  SDS
thermo fisher scientific
305000050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 102.13
Quantity 50g
Color Clear colorless
EINECS Number 214-323-1
RTECS Number JH2200000
Density 0.97g/mL
CAS Max % 100.0
Infrared Spectrum Authentic
Refractive Index 1.4140 to 1.4160 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
CAS Min % 98.5
Packaging Glass bottle
CAS 1120-97-4
Chemical Name or Material 4-Methyl-1, 3-dioxane
Molecular Formula C5H10O2
Assay Percent Range 99%
Name Note 99%
MDL Number MFCD00006569
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.