missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-1,3,2-dioxathiolane 2,2-Dioxide 98.0+%, TCI America™

$286.38 - $286.38

Chemical Identifiers

CAS 5689-83-8
Molecular Formula C3H6O4S
Molecular Weight (g/mol) 138.137
MDL Number MFCD08703327
InChI Key OQXNUCOGMMHHNA-UHFFFAOYSA-N
Synonym 1,2-Propanediol Cyclic Sulfate, 1,2-Propylene Sulfate, 4-Methyl-2,2-dioxo-1,3,2-dioxathiolane
PubChem CID 9964107
IUPAC Name 4-methyl-1,3,2-dioxathiolane 2,2-dioxide
SMILES CC1COS(=O)(=O)O1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M247025G View Documents Tci America
M247025G
25g Each for $286.38
 
Specifications

Chemical Identifiers

5689-83-8
138.137
OQXNUCOGMMHHNA-UHFFFAOYSA-N
9964107
CC1COS(=O)(=O)O1
C3H6O4S
MFCD08703327
1,2-Propanediol Cyclic Sulfate, 1,2-Propylene Sulfate, 4-Methyl-2,2-dioxo-1,3,2-dioxathiolane
4-methyl-1,3,2-dioxathiolane 2,2-dioxide

Specifications

50°C
25g
C3H6O4S
1,2-Propanediol Cyclic Sulfate, 1,2-Propylene Sulfate, 4-Methyl-2,2-dioxo-1,3,2-dioxathiolane
CC1COS(=O)(=O)O1
138.137
138.14
≥98.0% (GC)
Brown-Yellow
5689-83-8
MFCD08703327
OQXNUCOGMMHHNA-UHFFFAOYSA-N
4-methyl-1,3,2-dioxathiolane 2,2-dioxide
9964107
Clear Liquid at 20°C
4-Methyl-1,3,2-dioxathiolane 2,2-Dioxide
Safety and Handling

Safety and Handling

TSCA : No