Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-1,2-cyclohexanedicarboxylic anhydride, mixture of isomers, 98%

$53.04 - $144.20

Chemical Identifiers

Chemical Name or Material 4-Methyl-1,2-cyclohexanedicarboxylic anhydride
CAS 19438-60-9
Name Note mixture of isomers
Molecular Formula C9H12O3
Formula Weight 168.19
MDL Number MFCD00005927
Synonym Hexahydro-4-methylphthalic anhydride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2332822

  SDS
alfa aesar™
B2332822
100g Each for $53.04

AAB2332836

  SDS
alfa aesar™
B2332836
500g Each for $144.20
Description & Specifications

Specifications

Formula Weight 168.19
Chemical Name or Material 4-Methyl-1,2-cyclohexanedicarboxylic anhydride
TSCA TSCA
Molecular Formula C9H12O3
MDL Number MFCD00005927
CAS 19438-60-9
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P310-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Name Note mixture of isomers
Assay 98%
Synonym Hexahydro-4-methylphthalic anhydride
Health Hazard 1 Danger
Boiling Point 119° to 121°C (1mmHg)
Quantity 100g
Flash Point >110°C (230°F)
Odor Odorless
Health Hazard 2 GHS H Statement
H334-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P310-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.