Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methyl-1,2,3-thiadiazole-5-methanol, 99%

$60.27 - $149.92

Chemical Identifiers

Chemical Name or Material 4-Methyl-1,2,3-thiadiazole-5-methanol
CAS 163008-86-4
Molecular Formula C4H6N2OS
Formula Weight 130.17
MDL Number MFCD07371384
Synonym 5-Hydroxymethyl-4-methyl-1,2,3-thiadiazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2612903

  SDS
alfa aesar™
H2612903
1g Each for $149.92

AAH26129MD

  SDS
alfa aesar™
H26129MD
250mg Each for $60.27
Description & Specifications

Specifications

Formula Weight 130.17
Chemical Name or Material 4-Methyl-1,2,3-thiadiazole-5-methanol
Assay 99%
Molecular Formula C4H6N2OS
MDL Number MFCD07371384
CAS 163008-86-4
Synonym 5-Hydroxymethyl-4-methyl-1,2,3-thiadiazole
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.