Please login to your online account to display your discounted pricing

4-Methyl-1,2,3-thiadiazole-5-carboxylic acid, 97%, Maybridge


Quantity 25g

Manufacturer: fisher scientific company  CC02301EB

 View more versions of this product

Catalog No. CC02301EB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 144.15
Quantity 25g
Assay 97%
CAS Max % 100.0
CAS 18212-21-0
Molecular Formula C4H4N2O2S
CAS Min % 96.5
MDL Number MFCD00173830
Chemical Name or Material 4-Methyl-1, 2, 3-thiadiazole-5-carboxylic acid
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Methyl-1, 2, 3-thiadiazole-5-carboxylic acid
CAS 18212-21-0
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C4H4N2O2S
Formula Weight 144.15
MDL Number MFCD00173830
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.