missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-1,2,3,4-tetrahydroquinoline-6-carboxylic acid, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC452070010

 View more versions of this product

Catalog No. AC452070010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Methyl-1,2,3,4-tetrahydroquinoline-6-carboxylic acid
158°C to 160°C
1332627-30-1
C11H13NO2
QOQHRSQQPZYCGY-UHFFFAOYSA-N
4-methyl-1,2,3,4-tetrahydroquinoline-6-carboxylic acid
56766060
95%
Glass Bottle
1g
94% min. (HPLC)
1,2,3,4-tetrahydro-4-methylquinoline-6-carboxylic acid
CC1CCNC2=C1C=C(C=C2)C(=O)O
191.23
191.23

Chemical Identifiers

1332627-30-1
191.23
1,2,3,4-tetrahydro-4-methylquinoline-6-carboxylic acid
4-methyl-1,2,3,4-tetrahydroquinoline-6-carboxylic acid
C11H13NO2
QOQHRSQQPZYCGY-UHFFFAOYSA-N
56766060
CC1CCNC2=C1C=C(C=C2)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS Signal Word: Warning

SDS Documents