Learn More
4-Methyl-1,10-phenanthroline, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03188803
Description
4-methyl-1,10-phenanthroline can be used to produce 1,10-Phenanthrolin-4-aldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Methyl-1,10-phenanthroline | |
144°C to 145°C | |
MFCD00004978 | |
1,10-phenanthroline, 4-methyl, 4-methylpyridino 3,2-h quinoline, acmc-1aihj, 4-methyl-1,10 phenanthroline, 4-methyl-1,10-phenanthroline, 4-methyl-1,10-phenanthroline1,10-phenanthroline, 4-methyl | |
NAZZKEZTSOOCSZ-UHFFFAOYSA-N | |
4-methyl-1,10-phenanthroline | |
93149 | |
Crystalline |
31301-28-7 | |
C13H10N2 | |
1 g | |
Soluble in water. | |
CC1=C2C=CC3=C(C2=NC=C1)N=CC=C3 | |
194.237 | |
194.24 |
Chemical Identifiers
31301-28-7 | |
194.237 | |
NAZZKEZTSOOCSZ-UHFFFAOYSA-N | |
93149 | |
CC1=C2C=CC3=C(C2=NC=C1)N=CC=C3 |
C13H10N2 | |
MFCD00004978 | |
1,10-phenanthroline, 4-methyl, 4-methylpyridino 3,2-h quinoline, acmc-1aihj, 4-methyl-1,10 phenanthroline, 4-methyl-1,10-phenanthroline, 4-methyl-1,10-phenanthroline1,10-phenanthroline, 4-methyl | |
4-methyl-1,10-phenanthroline |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only