missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxytrityl Chloride [Hydroxyl Protecting Agent], TCI America™

Manufacturer:  TCI America M0790250G

 View more versions of this product

Catalog No. M0790250G



Specifications

Specifications

4-Methoxytrityl Chloride [Hydroxyl Protecting Agent]
124°C
14470-28-1
MFCD00000814
OBOHMJWDFPBPKD-UHFFFAOYSA-N
1-[chloro(diphenyl)methyl]-4-methoxybenzene
84462
Crystal-Powder at 20°C
Undesignated
250g
C20H17ClO
4-methoxytriphenylchloromethane, 4-methoxytrityl chloride, 4-methoxytriphenylmethyl chloride, 4-monomethoxytrityl chloride, mmtrcl, p-chlorodiphenylmethyl anisole, p-anisylchlorodiphenylmethane, 1-chlorodiphenylmethyl-4-methoxybenzene, chloro 4-methoxyphenyl methylene dibenzene, 4-methoxytritylchloride
COC1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Cl
308.805
308.81
≥98.0% (HPLC,T)

Chemical Identifiers

14470-28-1
308.805
OBOHMJWDFPBPKD-UHFFFAOYSA-N
84462
COC1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Cl
C20H17ClO
MFCD00000814
4-methoxytriphenylchloromethane, 4-methoxytrityl chloride, 4-methoxytriphenylmethyl chloride, 4-monomethoxytrityl chloride, mmtrcl, p-chlorodiphenylmethyl anisole, p-anisylchlorodiphenylmethane, 1-chlorodiphenylmethyl-4-methoxybenzene, chloro 4-methoxyphenyl methylene dibenzene, 4-methoxytritylchloride
1-[chloro(diphenyl)methyl]-4-methoxybenzene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents