missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxytrityl chloride, 97%

$56.92 - $875.92

Chemical Identifiers

CAS 14470-28-1
Molecular Formula C20H17ClO
Molecular Weight (g/mol) 308.805
MDL Number MFCD00000814
InChI Key OBOHMJWDFPBPKD-UHFFFAOYSA-N
Synonym 4-methoxytriphenylchloromethane, 4-methoxytrityl chloride, 4-methoxytriphenylmethyl chloride, 4-monomethoxytrityl chloride, mmtrcl, p-chlorodiphenylmethyl anisole, p-anisylchlorodiphenylmethane, 1-chlorodiphenylmethyl-4-methoxybenzene, chloro 4-methoxyphenyl methylene dibenzene, 4-methoxytritylchloride
PubChem CID 84462
IUPAC Name 1-[chloro(diphenyl)methyl]-4-methoxybenzene
SMILES COC1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1054509 View Documents Alfa Aesar™
A1054509
10g Each for $56.92
 
AAA1054530 View Documents Alfa Aesar™
A1054530
250g Each for $875.92
 
AAA1054518 View Documents Alfa Aesar™
A1054518
50g Each for $200.47
 
Description

Description

Applications
It is used as selective protecting reagent that is used for primary hydroxyl groups.

Solubility
Soluble in toluene.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

14470-28-1
308.805
OBOHMJWDFPBPKD-UHFFFAOYSA-N
84462
COC1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Cl
C20H17ClO
MFCD00000814
4-methoxytriphenylchloromethane, 4-methoxytrityl chloride, 4-methoxytriphenylmethyl chloride, 4-monomethoxytrityl chloride, mmtrcl, p-chlorodiphenylmethyl anisole, p-anisylchlorodiphenylmethane, 1-chlorodiphenylmethyl-4-methoxybenzene, chloro 4-methoxyphenyl methylene dibenzene, 4-methoxytritylchloride
1-[chloro(diphenyl)methyl]-4-methoxybenzene

Specifications

121°C to 125°C
14470-28-1
MFCD00000814
4-methoxytriphenylchloromethane, 4-methoxytrityl chloride, 4-methoxytriphenylmethyl chloride, 4-monomethoxytrityl chloride, mmtrcl, p-chlorodiphenylmethyl anisole, p-anisylchlorodiphenylmethane, 1-chlorodiphenylmethyl-4-methoxybenzene, chloro 4-methoxyphenyl methylene dibenzene, 4-methoxytritylchloride
COC1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Cl
308.805
308.81
4-Methoxytrityl chloride
10g
C20H17ClO
798425
OBOHMJWDFPBPKD-UHFFFAOYSA-N
1-[chloro(diphenyl)methyl]-4-methoxybenzene
84462
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning