missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxytoluene 98.0+%, TCI America™

Manufacturer:  Tci America M0149100ML

 View more versions of this product

Catalog No. M0149100ML



Specifications

Specifications

4-Methoxytoluene
Yellow
100mL
C8H10O
1993
CHLICZRVGGXEOD-UHFFFAOYSA-N
1-methoxy-4-methylbenzene
7731
Clear Liquid at 20°C
175°C
-50°C
104-93-8
MFCD00008413
4-methoxytoluene, 4-methylanisole, anisole, p-methyl, benzene, 1-methoxy-4-methyl, methyl p-tolyl ether, p-cresol methyl ether, p-cresyl methyl ether, p-methoxytoluene, p-methylanisole, p-tolyl methyl ether
CC1=CC=C(C=C1)OC
122.167
122.17
≥98.0% (GC)

Chemical Identifiers

104-93-8
122.167
CHLICZRVGGXEOD-UHFFFAOYSA-N
7731
CC1=CC=C(C=C1)OC
C8H10O
MFCD00008413
4-methoxytoluene, 4-methylanisole, anisole, p-methyl, benzene, 1-methoxy-4-methyl, methyl p-tolyl ether, p-cresol methyl ether, p-cresyl methyl ether, p-methoxytoluene, p-methylanisole, p-tolyl methyl ether
1-methoxy-4-methylbenzene
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0573&(3)-1035

RTECSNumber : BZ8780000

TSCA : Yes

Documents