missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxystyrene (stabilized with TBC), TCI America™

Manufacturer:  TCI America M013010ML

 View more versions of this product

Catalog No. M013010ML



Specifications

Specifications

4-Methoxystyrene (stabilized with TBC)
Undesignated
637-69-4
MFCD00008619
UAJRSHJHFRVGMG-UHFFFAOYSA-N
1-ethenyl-4-methoxybenzene
12507
Clear Liquid at 20°C
98°C
10mL
C9H10O
4-methoxystyrene, 1-methoxy-4-vinylbenzene, p-methoxystyrene, 4-vinylanisole, benzene, 1-ethenyl-4-methoxy, p-vinylanisole, anisole, p-vinyl, unii-2ish8t4a6e, ccris 4381, 4-vinylanisol
COC1=CC=C(C=C1)C=C
134.178
134.18
≥98.0% (GC)

Chemical Identifiers

637-69-4
134.178
UAJRSHJHFRVGMG-UHFFFAOYSA-N
12507
COC1=CC=C(C=C1)C=C
C9H10O
MFCD00008619
4-methoxystyrene, 1-methoxy-4-vinylbenzene, p-methoxystyrene, 4-vinylanisole, benzene, 1-ethenyl-4-methoxy, p-vinylanisole, anisole, p-vinyl, unii-2ish8t4a6e, ccris 4381, 4-vinylanisol
1-ethenyl-4-methoxybenzene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents