missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methoxysalicylic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 174240100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Methoxysalicylic acid | |
2237-36-7 | |
100.0 | |
White to Beige | |
98.5% min. (HPLC) | |
C8H8O4 | |
MFCD00002450 | |
10, 379 | |
MRIXVKKOHPQOFK-UHFFFAOYSA-N | |
2-hydroxy-4-methoxybenzoic acid | |
75231 | |
99% |
99% | |
98.5 | |
157.0°C to 159.0°C | |
Authentic | |
Glass bottle | |
CH3OC6H3(OH)CO2H | |
10 g | |
4-methoxysalicylic acid, 2-hydroxy-p-anisic acid, 2-hydroxy-4-methoxy-benzoic acid, benzoic acid, 2-hydroxy-4-methoxy, 2-hydroxy-4-methoxybenzoes, 4-methoxysalicylicacid, pubchem18895, 4-methoxy salicylic acid, 4-methoxy-salicylic acid, acmc-1cs6b | |
COC1=CC=C(C(O)=O)C(O)=C1 | |
168.15 | |
168.15 | |
Powder |
Chemical Identifiers
2237-36-7 | |
168.15 | |
MRIXVKKOHPQOFK-UHFFFAOYSA-N | |
75231 | |
COC1=CC=C(C(O)=O)C(O)=C1 |
C8H8O4 | |
MFCD00002450 | |
4-methoxysalicylic acid, 2-hydroxy-p-anisic acid, 2-hydroxy-4-methoxy-benzoic acid, benzoic acid, 2-hydroxy-4-methoxy, 2-hydroxy-4-methoxybenzoes, 4-methoxysalicylicacid, pubchem18895, 4-methoxy salicylic acid, 4-methoxy-salicylic acid, acmc-1cs6b | |
2-hydroxy-4-methoxybenzoic acid |
Safety and Handling
EINECSNumber : 218-801-0
RUO – Research Use Only