missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxysalicylamide, TCI America™

$229.17 - $799.05

Chemical Identifiers

CAS 6745-77-3
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD03840504
InChI Key BCYRZPCPWKCINY-UHFFFAOYSA-N
Synonym 2-Hydroxy-4-methoxybenzamide
PubChem CID 4868377
IUPAC Name 2-hydroxy-4-methoxybenzamide
SMILES COC1=CC(=C(C=C1)C(=O)N)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M18145G View Documents TCI America
M18145G
5g Each for $229.17
 
M181425G View Documents TCI America
M181425G
25g Each for $799.05
 
Specifications

Chemical Identifiers

6745-77-3
167.164
BCYRZPCPWKCINY-UHFFFAOYSA-N
4868377
COC1=CC(=C(C=C1)C(=O)N)O
C8H9NO3
MFCD03840504
2-Hydroxy-4-methoxybenzamide
2-hydroxy-4-methoxybenzamide

Specifications

Undesignated
5g
C8H9NO3
2-Hydroxy-4-methoxybenzamide
COC1=CC(=C(C=C1)C(=O)N)O
167.164
167.16
≥98.0% (HPLC,T)
158°C
6745-77-3
MFCD03840504
BCYRZPCPWKCINY-UHFFFAOYSA-N
2-hydroxy-4-methoxybenzamide
4868377
Crystal-Powder at 20°C
4-Methoxysalicylamide
Safety and Handling

Safety and Handling

TSCA : No