Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxypyridine N-oxide hydrate, 99%

$46.09 - $167.26

Chemical Identifiers

Chemical Name or Material 4-Methoxypyridine N-oxide hydrate
CAS 207511-18-0
Molecular Formula C6H7NO2·xH2O
Formula Weight 125.13 (Anhydrous)
MDL Number MFCD00149410
Beilstein 115212
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0470906

  SDS
alfa aesar™
L0470906
5g Each for $167.26

AAL0470903

  SDS
alfa aesar™
L0470903
1g Each for $46.09
Description & Specifications

Specifications

Formula Weight 125.13 (Anhydrous)
Molecular Formula C6H7NO2·xH2O
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
MDL Number MFCD00149410
Assay 99%
CAS 207511-18-0
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Beilstein 115212
Melting Point 78° to 82°C
Chemical Name or Material 4-Methoxypyridine N-oxide hydrate

Applications
4-Methoxypyridine N-oxide hydrate is used as an active pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.