missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxypyridine N-oxide, 98%

$92.84 - $350.07

Chemical Identifiers

CAS 1122-96-9
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.127
MDL Number MFCD00006207
InChI Key BOFAIBPJCWFJFT-UHFFFAOYSA-N
Synonym 4-methoxypyridine n-oxide, 4-methoxypyridine 1-oxide, 4-methoxypyridine-n-oxide, pyridine, 4-methoxy-, 1-oxide, 4-methoxypyridin-1-ium-1-olate, 4-methoxypyridine-n-oxide hydrate, pubchem2587, acmc-1buuj, 4-methoxypyridinen-oxide, 4-methoxypiridine-n-oxide
PubChem CID 70743
IUPAC Name 4-methoxy-1-oxidopyridin-1-ium
SMILES COC1=CC=[N+](C=C1)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1783003 View Documents Alfa Aesar™
L1783003
1g Each for $92.84
 
AAL1783006 View Documents Alfa Aesar™
L1783006
5g Each for $350.07
 
Specifications

Chemical Identifiers

1122-96-9
125.127
BOFAIBPJCWFJFT-UHFFFAOYSA-N
70743
COC1=CC=[N+](C=C1)[O-]
C6H7NO2
MFCD00006207
4-methoxypyridine n-oxide, 4-methoxypyridine 1-oxide, 4-methoxypyridine-n-oxide, pyridine, 4-methoxy-, 1-oxide, 4-methoxypyridin-1-ium-1-olate, 4-methoxypyridine-n-oxide hydrate, pubchem2587, acmc-1buuj, 4-methoxypyridinen-oxide, 4-methoxypiridine-n-oxide
4-methoxy-1-oxidopyridin-1-ium

Specifications

99°C to 104°C
1122-96-9
MFCD00006207
4-methoxypyridine n-oxide, 4-methoxypyridine 1-oxide, 4-methoxypyridine-n-oxide, pyridine, 4-methoxy-, 1-oxide, 4-methoxypyridin-1-ium-1-olate, 4-methoxypyridine-n-oxide hydrate, pubchem2587, acmc-1buuj, 4-methoxypyridinen-oxide, 4-methoxypiridine-n-oxide
COC1=CC=[N+](C=C1)[O-]
125.127
125.13
4-Methoxypyridine N-oxide
1g
C6H7NO2
115212
BOFAIBPJCWFJFT-UHFFFAOYSA-N
4-methoxy-1-oxidopyridin-1-ium
70743
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning