Please login to your online account to display your discounted pricing

4-Methoxypyridine, 99%, ACROS Organics™

$102.99 - $102.99

Chemical Identifiers

Chemical Name or Material 4-Methoxypyridine
CAS 620-08-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H7NO
Formula Weight 109.13
MDL Number MFCD00674049
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC379940050

  SDS
thermo fisher scientific
AC379940050
Glass bottle 5mL Each for $102.99

AC379940250

  SDS
fisher scientific acros
AC379940250
Glass bottle 25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 109.13
Physical Form Liquid
Quantity 5mL
Color Clear colorless to slightly yellow
Density 1.0750g/mL
CAS Max % 100.0
Beilstein 21, V,02, 98
Chemical Name or Material 4-Methoxypyridine
MDL Number MFCD00674049
Assay Percent Range 99%
Health Hazard 2 GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
CAS Min % 98.5
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5165 to 1.5215 (20°C, 589nm)
Storage Information Darkens on storage
Health Hazard 1 Warning
Molecular Formula C6H7NO
CAS 620-08-6
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.