missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4′-Methoxypropiophenone, TCI America™

$99.31 - $550.02

Chemical Identifiers

CAS 121-97-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00009310
InChI Key ZJVAWPKTWVFKHG-UHFFFAOYSA-N
Synonym 4'-methoxypropiophenone, p-methoxypropiophenone, 4-methoxypropiophenone, 1-4-methoxyphenyl propan-1-one, 1-propanone, 1-4-methoxyphenyl, propiophenone, 4'-methoxy, ethyl 4-methoxyphenyl ketone, 1-4-methoxyphenyl-1-propanone, 1-propanone,1-4-methoxyphenyl, 1-4-methoxy-phenyl-propan-1-one
PubChem CID 67144
IUPAC Name 1-(4-methoxyphenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(C=C1)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M171925G View Documents TCI America
M171925G
25g Each for $99.31
 
M1719250G View Documents TCI America
M1719250G
250g Each for $550.02
 
Specifications

Chemical Identifiers

121-97-1
164.204
ZJVAWPKTWVFKHG-UHFFFAOYSA-N
67144
CCC(=O)C1=CC=C(C=C1)OC
C10H12O2
MFCD00009310
4'-methoxypropiophenone, p-methoxypropiophenone, 4-methoxypropiophenone, 1-4-methoxyphenyl propan-1-one, 1-propanone, 1-4-methoxyphenyl, propiophenone, 4'-methoxy, ethyl 4-methoxyphenyl ketone, 1-4-methoxyphenyl-1-propanone, 1-propanone,1-4-methoxyphenyl, 1-4-methoxy-phenyl-propan-1-one
1-(4-methoxyphenyl)propan-1-one

Specifications

275°C
25g
C10H12O2
1325
ZJVAWPKTWVFKHG-UHFFFAOYSA-N
1-(4-methoxyphenyl)propan-1-one
67144
Crystal at 20°C
4′-Methoxypropiophenone
White-Yellow
121-97-1
MFCD00009310
4'-methoxypropiophenone, p-methoxypropiophenone, 4-methoxypropiophenone, 1-4-methoxyphenyl propan-1-one, 1-propanone, 1-4-methoxyphenyl, propiophenone, 4'-methoxy, ethyl 4-methoxyphenyl ketone, 1-4-methoxyphenyl-1-propanone, 1-propanone,1-4-methoxyphenyl, 1-4-methoxy-phenyl-propan-1-one
CCC(=O)C1=CC=C(C=C1)OC
164.204
164.20
≥97.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1246

TSCA : Yes