missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxyphthalic Acid, TCI America™

$96.25 - $296.39

Chemical Identifiers

CAS 1885-13-8
Molecular Formula C9H8O5
Molecular Weight (g/mol) 196.158
MDL Number MFCD00094739
InChI Key JKZSIEDAEHZAHQ-UHFFFAOYSA-N
PubChem CID 74665
IUPAC Name 4-methoxyphthalic acid
SMILES COC1=CC(=C(C=C1)C(=O)O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M14321G View Documents TCI America
M14321G
1g Each for $96.25
 
M14325G View Documents TCI America
M14325G
5g Each for $296.39
 
Specifications

Chemical Identifiers

1885-13-8
196.158
JKZSIEDAEHZAHQ-UHFFFAOYSA-N
4-methoxyphthalic acid
C9H8O5
MFCD00094739
74665
COC1=CC(=C(C=C1)C(=O)O)C(=O)O

Specifications

White-Yellow
1885-13-8
MFCD00094739
COC1=CC(=C(C=C1)C(=O)O)C(=O)O
196.158
196.16
≥98.0% (GC,T)
1g
C9H8O5
JKZSIEDAEHZAHQ-UHFFFAOYSA-N
4-methoxyphthalic acid
74665
Crystal-Powder at 20°C
4-Methoxyphthalic Acid
Safety and Handling

Safety and Handling

TSCA : No