Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxyphenylurea, 98+%

$47.32 - $47.32

Chemical Identifiers

Chemical Name or Material 4-Methoxyphenylurea
CAS 1566-42-3
Molecular Formula C8H10N2O2
Formula Weight 166.18
MDL Number MFCD00014789
Beilstein 2803938
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0120306

  SDS
alfa aesar™
L0120306
5g Each for $47.32
Description & Specifications

Specifications

Formula Weight 166.18
Chemical Name or Material 4-Methoxyphenylurea
Assay ≥98%
MDL Number MFCD00014789
CAS 1566-42-3
Quantity 5g
Beilstein 2803938
RTECS Number YT6451200
Melting Point 165° to 168°C
Molecular Formula C8H10N2O2