missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxyphenylphosphonic Dichloride, TCI America™

$88.61 - $291.81

Chemical Identifiers

CAS 37632-18-1
Molecular Formula C7H7Cl2O2P
Molecular Weight (g/mol) 225.005
MDL Number MFCD00123296
InChI Key LOFIFCKKPQFWSH-UHFFFAOYSA-N
Synonym 4-Dichlorophosphonylanisole
PubChem CID 216711
IUPAC Name 1-dichlorophosphoryl-4-methoxybenzene
SMILES COC1=CC=C(C=C1)P(=O)(Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M11565G View Documents TCI America
M11565G
5g Each for $291.81
 
M11561G View Documents TCI America
M11561G
1g Each for $88.61
 
Specifications

Chemical Identifiers

37632-18-1
225.005
LOFIFCKKPQFWSH-UHFFFAOYSA-N
216711
COC1=CC=C(C=C1)P(=O)(Cl)Cl
C7H7Cl2O2P
MFCD00123296
4-Dichlorophosphonylanisole
1-dichlorophosphoryl-4-methoxybenzene

Specifications

Undesignated
37632-18-1
MFCD00123296
4-Dichlorophosphonylanisole
COC1=CC=C(C=C1)P(=O)(Cl)Cl
225.005
225.00
≥90.0% (GC)
5g
C7H7Cl2O2P
3265
LOFIFCKKPQFWSH-UHFFFAOYSA-N
1-dichlorophosphoryl-4-methoxybenzene
216711
Clear Liquid at 20°C
4-Methoxyphenylphosphonic Dichloride
Safety and Handling

Safety and Handling

RTECSNumber : TA1825000

TSCA : No