missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxyphenylmagnesium bromide, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 377421000

 View more versions of this product

Catalog No. AC377421000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Methoxyphenylmagnesium bromide
19.02,74.64
Yellow to Brown
−21°C
C7H7BrMgO
MFCD00013515
1
Solubility in water: vigorous reaction
COC1=CC=C([Mg]Br)C=C1
211.34
211.34
13139-86-1,109-99-9
25.36,80.98
1.0000g/mL
AcroSeal™ Glass bottle
CH3O(C6H4)MgBr
100 mL
4-methoxyphenylmagnesium bromide, grignard reagent, bromo 4-methoxyphenyl magnesium, p-methoxyphenylmagnesium bromide, p-methoxyphenylmagnesium bromide, 4-methoxyphenymagnesium bromide, 4-methoxyphenymagnesium bromide, p-methoxyphenyl magnesium bromide, p-methoxyphenyl magnesium bromide, 4-methoxy-phenyl-magnesiumbromide, 4-methoxy-phenyl-magnesiumbromide
JKXAZEXIOPFJSC-UHFFFAOYSA-M
magnesium;methoxybenzene;bromide
2734898
Liquid

Chemical Identifiers

13139-86-1
211.34
JKXAZEXIOPFJSC-UHFFFAOYSA-M
2734898
COC1=CC=C([Mg]Br)C=C1
C7H7BrMgO
MFCD00013515
4-methoxyphenylmagnesium bromide, grignard reagent, bromo 4-methoxyphenyl magnesium, p-methoxyphenylmagnesium bromide, p-methoxyphenylmagnesium bromide, 4-methoxyphenymagnesium bromide, 4-methoxyphenymagnesium bromide, p-methoxyphenyl magnesium bromide, p-methoxyphenyl magnesium bromide, 4-methoxy-phenyl-magnesiumbromide, 4-methoxy-phenyl-magnesiumbromide
magnesium;methoxybenzene;bromide
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

Recommended Storage : Magnesium salts may crystallize below 20°C

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only