missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxyphenylmagnesium bromide, 0.5M in THF

Manufacturer:  Alfa Aesar™ 89435AP

 View more versions of this product

Catalog No. AA89435AP



Specifications

Specifications

4-Methoxyphenylmagnesium bromide
Air & Moisture Sensitive
500mL
C7H7BrMgO
UN2924
0.5M in THF
COC1=CC=[C-]C=C1.[Mg+2].[Br-]
211.341
211.34
0.955g/mL
Brown
13139-86-1
MFCD00013515
4-methoxyphenylmagnesium bromide, 4-methoxyphenylmagnesium bromide solution, bromo 4-methoxyphenyl magnesium, p-methoxyphenylmagnesium bromide, 4-methoxyphenyl magnesium bromide, 4-methoxyphenymagnesium bromide, 4-methoxy phenylmagnesiumbromide, p-methoxyphenyl magnesium bromide, p-methoxyphenyl-magnesium bromide, 4-methoxy-phenyl-magnesiumbromide
RBWRWAUAVRMBAC-UHFFFAOYSA-M
magnesium;methoxybenzene;bromide
2734898
Liquid

Chemical Identifiers

13139-86-1
211.341
RBWRWAUAVRMBAC-UHFFFAOYSA-M
2734898
COC1=CC=[C-]C=C1.[Mg+2].[Br-]
C7H7BrMgO
MFCD00013515
4-methoxyphenylmagnesium bromide, 4-methoxyphenylmagnesium bromide solution, bromo 4-methoxyphenyl magnesium, p-methoxyphenylmagnesium bromide, 4-methoxyphenyl magnesium bromide, 4-methoxyphenymagnesium bromide, 4-methoxy phenylmagnesiumbromide, p-methoxyphenyl magnesium bromide, p-methoxyphenyl-magnesium bromide, 4-methoxy-phenyl-magnesiumbromide
magnesium;methoxybenzene;bromide
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H351-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

SDS Documents