missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxyphenylhydrazine hydrochloride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC166490010

 View more versions of this product

Catalog No. AC166490010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Methoxyphenylhydrazine hydrochloride
Glass bottle
153°C to 154°C
98%
97.5
98%
CH3OC6H4NHNH2·HCl
15, 597
Solubility in water: soluble
COC1=CC=C(C=C1)NN.Cl
174.63
174.63
98%
Authentic
Pink to Purple
1g
19501-58-7
100.0
C7H10N2O·ClH
MFCD00012945
1-4-methoxyphenyl hydrazine hydrochloride
FQHCPFMTXFJZJS-UHFFFAOYSA-N
(4-methoxyphenyl)hydrazine;hydrochloride
2723904
Powder, Crystals and/or Chunks

Chemical Identifiers

19501-58-7
174.63
FQHCPFMTXFJZJS-UHFFFAOYSA-N
2723904
COC1=CC=C(C=C1)NN.Cl
C7H10N2O·ClH
MFCD00012945
1-4-methoxyphenyl hydrazine hydrochloride
(4-methoxyphenyl)hydrazine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Call a POISON CENTER or doctor/physician if

GHS Signal Word: Warning

EINECSNumber : 243-115-3

SDS Documents