missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxyphenylglyoxal hydrate, 95%, dry wt. basis, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L2027403

 View more versions of this product

Catalog No. AAL2027403



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Methoxyphenylglyoxal hydrate
104°C to 111°C
(dry wt.)
MFCD00234534
606701
NLUFPYPVEHQAQN-UHFFFAOYSA-N
2-(4-methoxyphenyl)-2-oxoacetaldehyde;hydrate
15556730
95%
16208-17-6
145°C to 147°C (13 mmHg)
C9H10O4
1 g
4-methoxyphenylglyoxal hydrate, 2-4-methoxyphenyl-2-oxoacetaldehyde hydrate, pubchem12129, acmc-20ai17, 4-methoxyphenyl glyoxal hydrate, p-methoxyphenylglyoxal monohydrate, 2-4-methoxyphenyl-2-oxoethanal, hydrate, ethanone,2,2-dihydroxy-1-4-methoxyphenyl, 4-methoxyphenyl oxo acetaldehyde-water 1/1, 2-4-methoxyphenyl-2-oxidanylidene-ethanal hydrate
COC1=CC=C(C=C1)C(=O)C(O)O
182.18
164.16 (Anhydrous)

Chemical Identifiers

16208-17-6
182.18
NLUFPYPVEHQAQN-UHFFFAOYSA-N
15556730
COC1=CC=C(C=C1)C(=O)C(O)O
C9H10O4
MFCD00234534
4-methoxyphenylglyoxal hydrate, 2-4-methoxyphenyl-2-oxoacetaldehyde hydrate, pubchem12129, acmc-20ai17, 4-methoxyphenyl glyoxal hydrate, p-methoxyphenylglyoxal monohydrate, 2-4-methoxyphenyl-2-oxoethanal, hydrate, ethanone,2,2-dihydroxy-1-4-methoxyphenyl, 4-methoxyphenyl oxo acetaldehyde-water 1/1, 2-4-methoxyphenyl-2-oxidanylidene-ethanal hydrate
2-(4-methoxyphenyl)-2-oxoacetaldehyde;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only