missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxyphenylacetyl chloride, 98%, Thermo Scientific™

Chemical Identifiers

CAS 4693-91-8
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.62
MDL Number MFCD00075474
InChI Key CXJOONIFSVSFAD-UHFFFAOYSA-N
Synonym 4-methoxyphenylacetyl chloride, 2-4-methoxyphenyl acetyl chloride, benzenacetyl chloride, 4-methoxy, benzeneacetyl chloride, 4-methoxy, 4-methoxyphenylacetylchloride, p-methoxyphenyl acetyl chloride, p-methoxyphenylacetyl chloride, acmc-1aso1, 4-methoxyphenylacetylchlorid, 4-methoxybenzeneacetyl chloride
PubChem CID 2734688
IUPAC Name 2-(4-methoxyphenyl)acetyl chloride
SMILES COC1=CC=C(C=C1)CC(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC368930020
SDS View Documents Promotion Details
Thermo Scientific Chemicals
368930020
2g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC368930100
SDS View Documents Promotion Details
Thermo Scientific Chemicals
368930100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

4693-91-8
184.62
CXJOONIFSVSFAD-UHFFFAOYSA-N
2734688
COC1=CC=C(C=C1)CC(=O)Cl
C9H9ClO2
MFCD00075474
4-methoxyphenylacetyl chloride, 2-4-methoxyphenyl acetyl chloride, benzenacetyl chloride, 4-methoxy, benzeneacetyl chloride, 4-methoxy, 4-methoxyphenylacetylchloride, p-methoxyphenyl acetyl chloride, p-methoxyphenylacetyl chloride, acmc-1aso1, 4-methoxyphenylacetylchlorid, 4-methoxybenzeneacetyl chloride
2-(4-methoxyphenyl)acetyl chloride

Specifications

4693-91-8
100.0
143.0°C (10.0 mmHg)
97.5% min. (GC)
C9H9ClO2
MFCD00075474
Glass bottle
2g
Solubility in water: reacts
CXJOONIFSVSFAD-UHFFFAOYSA-N
2-(4-methoxyphenyl)acetyl chloride
2734688
Liquid
4-Methoxyphenylacetyl chloride
97.5
1.2080g/mL
>110°C
Authentic
CH3OC6H4CH2COCl
10, III, 434
1.5390 to 1.5410
4-methoxyphenylacetyl chloride, 2-4-methoxyphenyl acetyl chloride, benzenacetyl chloride, 4-methoxy, benzeneacetyl chloride, 4-methoxy, 4-methoxyphenylacetylchloride, p-methoxyphenyl acetyl chloride, p-methoxyphenylacetyl chloride, acmc-1aso1, 4-methoxyphenylacetylchlorid, 4-methoxybenzeneacetyl chloride
1.208
COC1=CC=C(C=C1)CC(=O)Cl
184.62
184.62
98%
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

SDS

RUO – Research Use Only