missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methoxyphenylacetonitrile, 98%, Thermo Scientific Chemicals
$54.12 - $140.96
Chemical Identifiers
CAS | 104-47-2 |
---|---|
Molecular Formula | C9H9NO |
Molecular Weight (g/mol) | 147.177 |
MDL Number | MFCD00001919 |
InChI Key | PACGLQCRGWFBJH-UHFFFAOYSA-N |
Synonym | 4-methoxyphenylacetonitrile, 2-4-methoxyphenyl acetonitrile, 4-methoxybenzyl cyanide, 4-methoxyphenyl acetonitrile, p-methoxyphenylacetonitrile, benzeneacetonitrile, 4-methoxy, p-methoxybenzyl cyanide, anisylacetonitrile, 4-anisyl cyanide, p-methoxybenzeneacetonitrile |
PubChem CID | 66031 |
IUPAC Name | 2-(4-methoxyphenyl)acetonitrile |
SMILES | COC1=CC=C(C=C1)CC#N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1505614
|
Thermo Scientific Chemicals
A1505614 |
25 g |
Each for $54.12
|
|
|||||
AAA1505622
|
Thermo Scientific Chemicals
A1505622 |
100 g |
Each for $140.96
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
104-47-2 | |
147.177 | |
PACGLQCRGWFBJH-UHFFFAOYSA-N | |
66031 | |
COC1=CC=C(C=C1)CC#N |
C9H9NO | |
MFCD00001919 | |
4-methoxyphenylacetonitrile, 2-4-methoxyphenyl acetonitrile, 4-methoxybenzyl cyanide, 4-methoxyphenyl acetonitrile, p-methoxyphenylacetonitrile, benzeneacetonitrile, 4-methoxy, p-methoxybenzyl cyanide, anisylacetonitrile, 4-anisyl cyanide, p-methoxybenzeneacetonitrile | |
2-(4-methoxyphenyl)acetonitrile |
Specifications
104-47-2 | |
1.085 | |
117°C (242°F) | |
1.53 | |
25 g | |
509162 | |
PACGLQCRGWFBJH-UHFFFAOYSA-N | |
2-(4-methoxyphenyl)acetonitrile | |
66031 | |
98% |
8°C | |
285°C to 287°C | |
C9H9NO | |
MFCD00001919 | |
UN3276 | |
4-methoxyphenylacetonitrile, 2-4-methoxyphenyl acetonitrile, 4-methoxybenzyl cyanide, 4-methoxyphenyl acetonitrile, p-methoxyphenylacetonitrile, benzeneacetonitrile, 4-methoxy, p-methoxybenzyl cyanide, anisylacetonitrile, 4-anisyl cyanide, p-methoxybenzeneacetonitrile | |
COC1=CC=C(C=C1)CC#N | |
147.177 | |
147.18 | |
4-Methoxyphenylacetonitrile |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber : 203-206-0
RTECSNumber : AM0810000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only