missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxyphenylacetone 97.0+%, TCI America™
Supplier: TCI America M0675100ML
Specifications
| 4-Methoxyphenylacetone | |
| Yellow | |
| C10H12O2 | |
| 100 mL | |
| WFWKNGZODAOLEO-UHFFFAOYSA-N | |
| 1-(4-methoxyphenyl)propan-2-one | |
| 31231 | |
| ≥97.0% (GC) |
| 122-84-9 | |
| 269°C | |
| MFCD00008773 | |
| 4-methoxyphenylacetone, p-acetonylanisole, anisketone, 1-4-methoxyphenyl propan-2-one, 2-propanone, 1-4-methoxyphenyl, anisyl methyl ketone, p-methoxyphenylacetone, 4-methoxybenzyl methyl ketone, 1-p-methoxyphenyl-2-propanone, anisic ketone | |
| CC(=O)CC1=CC=C(C=C1)OC | |
| 164.204 | |
| 164.20 | |
| Liquid |
Chemical Identifiers
| 122-84-9 | |
| 164.204 | |
| WFWKNGZODAOLEO-UHFFFAOYSA-N | |
| 31231 | |
| CC(=O)CC1=CC=C(C=C1)OC |
| C10H12O2 | |
| MFCD00008773 | |
| 4-methoxyphenylacetone, p-acetonylanisole, anisketone, 1-4-methoxyphenyl propan-2-one, 2-propanone, 1-4-methoxyphenyl, anisyl methyl ketone, p-methoxyphenylacetone, 4-methoxybenzyl methyl ketone, 1-p-methoxyphenyl-2-propanone, anisic ketone | |
| 1-(4-methoxyphenyl)propan-2-one |
Safety and Handling
RTECSNumber : UC2990000
TSCA : Yes