missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methoxyphenylacetone, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 189800050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Methoxyphenylacetone | |
122-84-9 | |
100.0 | |
1.0670g/mL | |
135°C | |
98% min. (GC) | |
C10H12O2 | |
CH3OC6H4CH2COCH3 | |
5 g | |
1.067 | |
WFWKNGZODAOLEO-UHFFFAOYSA-N | |
1-(4-methoxyphenyl)propan-2-one | |
31231 | |
98+% |
98+% | |
98.0 | |
Green-Yellow to Yellow | |
266°C to 268°C | |
Authentic | |
Glass bottle | |
1.5200 to 1.53 | |
MFCD00008773 | |
08, IV, 460 | |
4-methoxyphenylacetone, p-acetonylanisole, anisketone, 1-4-methoxyphenyl propan-2-one, 2-propanone, 1-4-methoxyphenyl, anisyl methyl ketone, p-methoxyphenylacetone, 4-methoxybenzyl methyl ketone, 1-p-methoxyphenyl-2-propanone, anisic ketone | |
CC(=O)CC1=CC=C(C=C1)OC | |
164.2 | |
164.2 | |
Liquid |
Chemical Identifiers
122-84-9 | |
164.2 | |
WFWKNGZODAOLEO-UHFFFAOYSA-N | |
31231 | |
CC(=O)CC1=CC=C(C=C1)OC |
C10H12O2 | |
MFCD00008773 | |
4-methoxyphenylacetone, p-acetonylanisole, anisketone, 1-4-methoxyphenyl propan-2-one, 2-propanone, 1-4-methoxyphenyl, anisyl methyl ketone, p-methoxyphenylacetone, 4-methoxybenzyl methyl ketone, 1-p-methoxyphenyl-2-propanone, anisic ketone | |
1-(4-methoxyphenyl)propan-2-one |
Safety and Handling
EINECSNumber : 204-578-7
RTECSNumber : UC2990000
TSCA : TSCA
RUO – Research Use Only