missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methoxyphenyl trans-4-n-pentylcyclohexanecarboxylate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H5579203
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications4-Methoxyphenyl trans-4-n-pentylcyclohexanecarboxylate, is mainly used as a nematic liquid crystal monomer, liquid crystal material. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Sparingly Soluble in water (1.7E-3 g/L) (25°C).
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
4-Methoxyphenyl trans-4-n-pentylcyclohexanecarboxylate | |
40°C to 44°C | |
MFCD11053432 | |
cyclohexanecarboxylic acid, 4-pentyl-, 4-methoxyphenyl ester, trans, trans-4-methoxyphenyl 4-pentylcyclohexanecarboxylate, trans-4-methoxy-phenyl 4-pentyl-cyclohexanecarboxylate, trans-4-pentylcyclohexanecarboxylic acid, 4-methoxyphenyl ester, 4-methoxyphenyl trans-4-pentylcyclohexanoate, 4-methoxyphenyl 4-pentylcyclohexanecarboxylate, 4-methoxyphenyl trans-4-pentylcyclohexanecarboxylate, hjivfzuptyadsp-wkilwmfisa-n, 4-methoxyphenyl 4-pentylcyclohexanecarboxylate #, 4-methoxyphenyl 4-pentylcyclohexane-1-carboxylate | |
HJIVFZUPTYADSP-UHFFFAOYSA-N | |
(4-methoxyphenyl) 4-pentylcyclohexane-1-carboxylate | |
105460 | |
97% |
67589-52-0 | |
C19H28O3 | |
1 g | |
Sparingly Soluble in water (1.7E-3g/L) (25°C). | |
CCCCCC1CCC(CC1)C(=O)OC2=CC=C(C=C2)OC | |
304.43 | |
304.43 |
Chemical Identifiers
67589-52-0 | |
304.43 | |
HJIVFZUPTYADSP-UHFFFAOYSA-N | |
105460 | |
CCCCCC1CCC(CC1)C(=O)OC2=CC=C(C=C2)OC |
C19H28O3 | |
MFCD11053432 | |
cyclohexanecarboxylic acid, 4-pentyl-, 4-methoxyphenyl ester, trans, trans-4-methoxyphenyl 4-pentylcyclohexanecarboxylate, trans-4-methoxy-phenyl 4-pentyl-cyclohexanecarboxylate, trans-4-pentylcyclohexanecarboxylic acid, 4-methoxyphenyl ester, 4-methoxyphenyl trans-4-pentylcyclohexanoate, 4-methoxyphenyl 4-pentylcyclohexanecarboxylate, 4-methoxyphenyl trans-4-pentylcyclohexanecarboxylate, hjivfzuptyadsp-wkilwmfisa-n, 4-methoxyphenyl 4-pentylcyclohexanecarboxylate #, 4-methoxyphenyl 4-pentylcyclohexane-1-carboxylate | |
(4-methoxyphenyl) 4-pentylcyclohexane-1-carboxylate |
Safety and Handling
EINECSNumber : 266-746-6
TSCA : Yes
Recommended Storage : Ambient temperatures