missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxyphenyl Isothiocyanate 98.0+%, TCI America™

$87.23 - $87.23

Chemical Identifiers

CAS 2284-20-0
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00011676
InChI Key VRPQCVLBOZOYCG-UHFFFAOYSA-N
Synonym 4-methoxyphenyl isothiocyanate, 4-methoxyphenylisothiocyanate, 1-isothiocyanato-4-methoxy-benzene, p-methoxyphenyl isothiocyanate, benzene, 1-isothiocyanato-4-methoxy, isothiocyanic acid 4-methoxyphenyl ester, 4-methoxybenzenisothiocyanate, 4-isothiocyanatoanisole, acmc-1ccqx, p-methoxyphenylisothiocyanate
PubChem CID 75293
IUPAC Name 1-isothiocyanato-4-methoxybenzene
SMILES COC1=CC=C(C=C1)N=C=S
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I05135G View Documents Tci America
I05135G
5g Each for $87.23
 
Specifications

Chemical Identifiers

2284-20-0
165.21
VRPQCVLBOZOYCG-UHFFFAOYSA-N
75293
COC1=CC=C(C=C1)N=C=S
C8H7NOS
MFCD00011676
4-methoxyphenyl isothiocyanate, 4-methoxyphenylisothiocyanate, 1-isothiocyanato-4-methoxy-benzene, p-methoxyphenyl isothiocyanate, benzene, 1-isothiocyanato-4-methoxy, isothiocyanic acid 4-methoxyphenyl ester, 4-methoxybenzenisothiocyanate, 4-isothiocyanatoanisole, acmc-1ccqx, p-methoxyphenylisothiocyanate
1-isothiocyanato-4-methoxybenzene

Specifications

281°C
18°C
2284-20-0
MFCD00011676
4-methoxyphenyl isothiocyanate, 4-methoxyphenylisothiocyanate, 1-isothiocyanato-4-methoxy-benzene, p-methoxyphenyl isothiocyanate, benzene, 1-isothiocyanato-4-methoxy, isothiocyanic acid 4-methoxyphenyl ester, 4-methoxybenzenisothiocyanate, 4-isothiocyanatoanisole, acmc-1ccqx, p-methoxyphenylisothiocyanate
COC1=CC=C(C=C1)N=C=S
165.21
165.21
≥98.0% (GC)
Yellow
5g
C8H7NOS
1760
VRPQCVLBOZOYCG-UHFFFAOYSA-N
1-isothiocyanato-4-methoxybenzene
75293
Clear Liquid at 20°C
4-Methoxyphenyl Isothiocyanate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator