missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxyphenyl beta-D-glucopyranoside, 98%

$99.47 - $547.45

Chemical Identifiers

CAS 6032-32-2
Molecular Formula C13H18O7
Molecular Weight (g/mol) 286.28
MDL Number MFCD06797143
InChI Key SIXFVXJMCGPTRB-UJPOAAIJSA-N
Synonym methylarbutin, 4-methoxyphenyl beta-d-glucopyranoside, p-methoxyphenyl b-d-glucoside, unii-f9mw5fu0xh, 4-methoxyphenyl b-d-glucopyranoside, f9mw5fu0xh, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-4-methoxyphenoxy oxane-3,4,5-triol, methylarbutoside, 4-methoxyphenylglucoside, p-methoxyphenyl beta-d-glucopyranoside
PubChem CID 80131
ChEBI CHEBI:81207
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxane-3,4,5-triol
SMILES COC1=CC=C(C=C1)OC2C(C(C(C(O2)CO)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6642203 View Documents Alfa Aesar™
J6642203
1g Each for $99.47
 
AAJ6642206 View Documents Alfa Aesar™
J6642206
5g Each for $547.45
 
Specifications

Chemical Identifiers

6032-32-2
286.28
SIXFVXJMCGPTRB-UJPOAAIJSA-N
80131
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxane-3,4,5-triol
C13H18O7
MFCD06797143
methylarbutin, 4-methoxyphenyl beta-d-glucopyranoside, p-methoxyphenyl b-d-glucoside, unii-f9mw5fu0xh, 4-methoxyphenyl b-d-glucopyranoside, f9mw5fu0xh, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-4-methoxyphenoxy oxane-3,4,5-triol, methylarbutoside, 4-methoxyphenylglucoside, p-methoxyphenyl beta-d-glucopyranoside
CHEBI:81207
COC1=CC=C(C=C1)OC2C(C(C(C(O2)CO)O)O)O

Specifications

−64.5° (c=1 in Water)
175°C to 179°C
6032-32-2
MFCD06797143
Soluble in DMSO or water
COC1=CC=C(C=C1)OC2C(C(C(C(O2)CO)O)O)O
286.28
CHEBI:81207
Powder
4-Methoxyphenyl beta-D-glucopyranoside
White
1g
C13H18O7
methylarbutin, 4-methoxyphenyl beta-d-glucopyranoside, p-methoxyphenyl b-d-glucoside, unii-f9mw5fu0xh, 4-methoxyphenyl b-d-glucopyranoside, f9mw5fu0xh, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-4-methoxyphenoxy oxane-3,4,5-triol, methylarbutoside, 4-methoxyphenylglucoside, p-methoxyphenyl beta-d-glucopyranoside
SIXFVXJMCGPTRB-UJPOAAIJSA-N
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxane-3,4,5-triol
80131
286.28
98%