missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxyphenyl beta-D-Galactopyranoside, TCI America™

$220.00 - $716.55

Chemical Identifiers

CAS 3150-20-7
Molecular Formula C13H18O7
Molecular Weight (g/mol) 286.28
MDL Number MFCD06797126
InChI Key SIXFVXJMCGPTRB-KSSYENDESA-N
PubChem CID 11300643
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxane-3,4,5-triol
SMILES COC1=CC=C(C=C1)OC2C(C(C(C(O2)CO)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M14815G View Documents TCI America
M14815G
5g Each for $220.00
 
M148125G View Documents TCI America
M148125G
25g Each for $716.55
 
Specifications

Chemical Identifiers

3150-20-7
286.28
SIXFVXJMCGPTRB-KSSYENDESA-N
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxane-3,4,5-triol
C13H18O7
MFCD06797126
11300643
COC1=CC=C(C=C1)OC2C(C(C(C(O2)CO)O)O)O

Specifications

White
5g
C13H18O7
SIXFVXJMCGPTRB-KSSYENDESA-N
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxane-3,4,5-triol
11300643
Crystal-Powder at 20°C
4-Methoxyphenyl beta-D-Galactopyranoside
160°C
3150-20-7
MFCD06797126
COC1=CC=C(C=C1)OC2C(C(C(C(O2)CO)O)O)O
286.28
286.28
≥98.0% (HPLC)
Safety and Handling

Safety and Handling

TSCA : No