Learn More
(4-Methoxyphenyl)acetonitrile, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 173760250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
(4-Methoxyphenyl)acetonitrile | |
96.0 | |
8.0°C | |
1.0800g/mL | |
117°C | |
96% min. (GC) | |
C9H9NO | |
CH3OC6H4CH2CN | |
25 g | |
1.08 | |
Solubility in water: 10g/L (20°C). Other solubilities: soluble in ethyl alcohol,ethyl ether,chloroform,miscible in isopropanol,acetone,toluene | |
COC1=CC=C(C=C1)CC#N | |
147.18 | |
147.18 | |
Liquid |
104-47-2 | |
100.0 | |
Colorless to Yellow | |
286.0°C to 287.0°C | |
Authentic | |
Glass bottle | |
1.53 | |
MFCD00001919 | |
10, 191 | |
4-methoxyphenylacetonitrile, 2-4-methoxyphenyl acetonitrile, 4-methoxybenzyl cyanide, 4-methoxyphenyl acetonitrile, p-methoxyphenylacetonitrile, benzeneacetonitrile, 4-methoxy, p-methoxybenzyl cyanide, anisylacetonitrile, 4-anisyl cyanide, p-methoxybenzeneacetonitrile | |
PACGLQCRGWFBJH-UHFFFAOYSA-N | |
2-(4-methoxyphenyl)acetonitrile | |
66031 | |
97% |
Chemical Identifiers
104-47-2 | |
PACGLQCRGWFBJH-UHFFFAOYSA-N | |
66031 | |
COC1=CC=C(C=C1)CC#N |
MFCD00001919 | |
4-methoxyphenylacetonitrile, 2-4-methoxyphenyl acetonitrile, 4-methoxybenzyl cyanide, 4-methoxyphenyl acetonitrile, p-methoxyphenylacetonitrile, benzeneacetonitrile, 4-methoxy, p-methoxybenzyl cyanide, anisylacetonitrile, 4-anisyl cyanide, p-methoxybenzeneacetonitrile | |
2-(4-methoxyphenyl)acetonitrile |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Rinse mouth.
Avoid r
GHS Signal Word: Danger
EINECSNumber : 203-206-
RUO – Research Use Only