Please login to your online account to display your discounted pricing

(4-Methoxyphenyl)(4-piperidyl)methanone hydrochloride, 97%, Maybridge


Quantity 250mg
Packaging Amber glass bottle

Manufacturer: fisher scientific company  SEW03999CB

 View more versions of this product

Catalog No. SEW03999CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 255.74
Quantity 250mg
Assay 97%
CAS Min % 97.0
Assay Percent Range 97%
CAS 25519-82-8
Molecular Formula C13H17NO2·ClH
MDL Number MFCD00114817
Packaging Amber glass bottle
Chemical Name or Material (4-Methoxyphenyl)(4-piperidyl)methanone hydrochloride
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

Specifications

Chemical Name or Material (4-Methoxyphenyl)(4-piperidyl)methanone hydrochloride
CAS 25519-82-8
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H17NO2·ClH
Formula Weight 255.74
MDL Number MFCD00114817
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.