missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxyphenyl 4-n-propylcyclohexanecarboxylate, 97%

Manufacturer:  Alfa Aesar™ H5506203

 View more versions of this product

Catalog No. AAH5506203



Specifications

Specifications

4-Methoxyphenyl 4-n-propylcyclohexanecarboxylate
67589-38-2
MFCD11053431
YOMOGIXKVWWEHG-UHFFFAOYSA-N
(4-methoxyphenyl) 4-propylcyclohexane-1-carboxylate
106715
97%
1g
C17H24O3
4-methoxyphenyl 1s,4r-4-propylcyclohexane-1-carboxylate
CCCC1CCC(CC1)C(=O)OC2=CC=C(C=C2)OC
276.376
276.38

Chemical Identifiers

67589-38-2
276.376
YOMOGIXKVWWEHG-UHFFFAOYSA-N
106715
CCCC1CCC(CC1)C(=O)OC2=CC=C(C=C2)OC
C17H24O3
MFCD11053431
4-methoxyphenyl 1s,4r-4-propylcyclohexane-1-carboxylate
(4-methoxyphenyl) 4-propylcyclohexane-1-carboxylate
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents