missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxyphenyl 3-O-Allyl-4,6-O-benzylidene-beta-D-galactopyranoside, TCI America™

$229.17 - $640.16

Chemical Identifiers

CAS 400091-05-6
Molecular Formula C23H26O7
Molecular Weight (g/mol) 414.454
MDL Number MFCD06797132
InChI Key BALLDQXJTPWKTQ-SFPLDMDMSA-N
PubChem CID 126970166
IUPAC Name (4aR,6R,7R,8R,8aS)-6-(4-methoxyphenoxy)-2-phenyl-8-prop-2-enoxy-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-7-ol
SMILES COC1=CC=C(C=C1)OC2C(C(C3C(O2)COC(O3)C4=CC=CC=C4)OCC=C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M15891G View Documents TCI America
M15891G
1g Each for $229.17
 
M15895G View Documents TCI America
M15895G
5g Each for $640.16
 
Specifications

Chemical Identifiers

400091-05-6
414.454
BALLDQXJTPWKTQ-SFPLDMDMSA-N
(4aR,6R,7R,8R,8aS)-6-(4-methoxyphenoxy)-2-phenyl-8-prop-2-enoxy-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-7-ol
C23H26O7
MFCD06797132
126970166
COC1=CC=C(C=C1)OC2C(C(C3C(O2)COC(O3)C4=CC=CC=C4)OCC=C)O

Specifications

White-Yellow
1g
C23H26O7
BALLDQXJTPWKTQ-SFPLDMDMSA-N
(4aR,6R,7R,8R,8aS)-6-(4-methoxyphenoxy)-2-phenyl-8-prop-2-enoxy-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-7-ol
126970166
Crystal-Powder at 20°C
4-Methoxyphenyl 3-O-Allyl-4,6-O-benzylidene-beta-D-galactopyranoside
228°C
400091-05-6
MFCD06797132
COC1=CC=C(C=C1)OC2C(C(C3C(O2)COC(O3)C4=CC=CC=C4)OCC=C)O
414.454
414.45
≥98.0% (HPLC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer