missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methoxyphenyl 2,3,4,6-Tetra-O-acetyl-beta-D-galactopyranoside, TCI America™
$152.78 - $436.95
Chemical Identifiers
CAS | 2872-65-3 |
---|---|
Molecular Formula | C21H26O11 |
Molecular Weight (g/mol) | 454.428 |
MDL Number | MFCD06797123 |
InChI Key | RPHXBVOPPUTUES-XDWAVFMPSA-N |
PubChem CID | 13325756 |
IUPAC Name | [(2R,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-(4-methoxyphenoxy)oxan-2-yl]methyl acetate |
SMILES | CC(=O)OCC1C(C(C(C(O1)OC2=CC=C(C=C2)OC)OC(=O)C)OC(=O)C)OC(=O)C |
Chemical Identifiers
2872-65-3 | |
454.428 | |
RPHXBVOPPUTUES-XDWAVFMPSA-N | |
[(2R,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-(4-methoxyphenoxy)oxan-2-yl]methyl acetate |
C21H26O11 | |
MFCD06797123 | |
13325756 | |
CC(=O)OCC1C(C(C(C(O1)OC2=CC=C(C=C2)OC)OC(=O)C)OC(=O)C)OC(=O)C |
Specifications
White-Yellow | |
5g | |
C21H26O11 | |
RPHXBVOPPUTUES-XDWAVFMPSA-N | |
[(2R,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-(4-methoxyphenoxy)oxan-2-yl]methyl acetate | |
13325756 | |
Crystal-Powder at 20°C | |
4-Methoxyphenyl 2,3,4,6-Tetra-O-acetyl-beta-D-galactopyranoside |
105°C | |
2872-65-3 | |
MFCD06797123 | |
CC(=O)OCC1C(C(C(C(O1)OC2=CC=C(C=C2)OC)OC(=O)C)OC(=O)C)OC(=O)C | |
454.428 | |
454.43 | |
≥98.0% (HPLC) |
Safety and Handling
TSCA : No
Recommended Storage : Freezer