missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxyphenol, TCI America™

Manufacturer:  TCI America M0123500G

 View more versions of this product

Catalog No. M0123500G



Specifications

Specifications

4-Methoxyphenol
Undesignated
500g
C7H8O2
mequinol, 4-hydroxyanisole, p-hydroxyanisole, p-methoxyphenol, hydroquinone monomethyl ether, phenol, 4-methoxy, leucobasal, mehq, leucodine b, mechinolum
COC1=CC=C(C=C1)O
124.139
CHEBI:69441
Crystal-Powder at 20°C
243°C
57°C
150-76-5
MFCD00002332
NWVVVBRKAWDGAB-UHFFFAOYSA-N
4-methoxyphenol
9015
124.14
≥99.0% (GC)

Chemical Identifiers

150-76-5
124.139
NWVVVBRKAWDGAB-UHFFFAOYSA-N
9015
4-methoxyphenol
C7H8O2
MFCD00002332
mequinol, 4-hydroxyanisole, p-hydroxyanisole, p-methoxyphenol, hydroquinone monomethyl ether, phenol, 4-methoxy, leucobasal, mehq, leucodine b, mechinolum
CHEBI:69441
COC1=CC=C(C=C1)O
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0567

RTECSNumber : SL7700000

TSCA : Yes

Documents