Learn More
4-Methoxyphenol, 98+%
CAS: 150-76-5 | C7H8O2 | 124.14 g/mol
Supplier: Thermo Scientific Chemicals A148450B
Description
4-Methoxyphenol is an active ingredient and used in dermatology. It is employed as a pharmaceutical drug in skin depigmentation. It is used as polymerization inhibitors. For example, in the radical polymerization of acryaltes and styrene monomers. It is also used as an intermediate in the preparationagrochemicals, liquid crystals. It acts as a stabilizer for the formulation of inks, toners and adhesives. It is mainly used as an additive for textile and leather industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methoxyphenol is an active ingredient and used in dermatology. It is employed as a pharmaceutical drug in skin depigmentation. It is used as polymerization inhibitors. For example, in the radical polymerization of acryaltes and styrene monomers. It is also used as an intermediate in the preparationagrochemicals, liquid crystals. It acts as a stabilizer for the formulation of inks, toners and adhesives. It is mainly used as an additive for textile and leather industries.
Solubility
Soluble in acetone, ethyl acetate, ethanol, ether, benzene and carbon tetrachloride.
Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed. Incompatible with halogens and oxidizing agents.
Specifications
| 4-Methoxyphenol | |
| 55°C to 58°C | |
| 243°C to 246°C | |
| C7H8O2 | |
| 1000 g | |
| mequinol, 4-hydroxyanisole, p-hydroxyanisole, p-methoxyphenol, hydroquinone monomethyl ether, phenol, 4-methoxy, leucobasal, mehq, leucodine b, mechinolum | |
| NWVVVBRKAWDGAB-UHFFFAOYSA-N | |
| 4-methoxyphenol | |
| 9015 | |
| 124.14 |
| 150-76-5 | |
| 1.55 | |
| 132°C (269°F) | |
| MFCD00002332 | |
| 507924 | |
| Soluble in acetone,ethyl acetate,ethanol,ether,benzene and carbon tetrachloride. | |
| COC1=CC=C(O)C=C1 | |
| 124.14 | |
| CHEBI:69441 | |
| ≥98% |
Chemical Identifiers
| 150-76-5 | |
| 124.14 | |
| NWVVVBRKAWDGAB-UHFFFAOYSA-N | |
| 9015 | |
| 4-methoxyphenol |
| C7H8O2 | |
| MFCD00002332 | |
| mequinol, 4-hydroxyanisole, p-hydroxyanisole, p-methoxyphenol, hydroquinone monomethyl ether, phenol, 4-methoxy, leucobasal, mehq, leucodine b, mechinolum | |
| CHEBI:69441 | |
| COC1=CC=C(O)C=C1 |
Safety and Handling
GHS H Statement
H302-H319-H317
Harmful if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P330-P333+P313-P363-P501c
H302-H317-H319-H361d-H373-H500
EINECSNumber : 205-769-8
RTECSNumber : SL7700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only