Learn More
4-Methoxynaphthalene-1-carboxylic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6443206
Description
Used in the preparation of α-aryl/pyridinyl ethanolamines as selective β3 adrenergic receptor agonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed in the preparation of α-aryl/pyridinyl ethanolamines as selective β3 adrenergic receptor agonists.
Solubility
Soluble in DMSO and chloroform. Slightly soluble in water.
Notes
Store away from acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
4-Methoxynaphthalene-1-carboxylic acid | |
243°C to 247°C | |
C12H10O3 | |
5 g | |
4-methoxy-1-naphthoic acid, 4-methoxy-naphthalene-1-carboxylic acid, 1-naphthalenecarboxylic acid, 4-methoxy, 1-naphthalenecarboxylicacid, 4-methoxy, 4-methoxynaphthalenecarboxylic acid, ksc181m4d, 4-methoxy-1-naphthalenecarboxylic acid | |
WRQHSQDGYYDRMX-UHFFFAOYSA-N | |
4-methoxynaphthalene-1-carboxylic acid | |
765380 | |
98% |
13041-62-8 | |
White | |
MFCD02575372 | |
2099800 | |
Soluble in DMSO and chloroform. Slightly soluble in water. | |
COC1=CC=C(C2=CC=CC=C21)C(=O)O | |
202.209 | |
202.21 | |
Crystalline Solid |
Chemical Identifiers
13041-62-8 | |
202.209 | |
WRQHSQDGYYDRMX-UHFFFAOYSA-N | |
765380 | |
COC1=CC=C(C2=CC=CC=C21)C(=O)O |
C12H10O3 | |
MFCD02575372 | |
4-methoxy-1-naphthoic acid, 4-methoxy-naphthalene-1-carboxylic acid, 1-naphthalenecarboxylic acid, 4-methoxy, 1-naphthalenecarboxylicacid, 4-methoxy, 4-methoxynaphthalenecarboxylic acid, ksc181m4d, 4-methoxy-1-naphthalenecarboxylic acid | |
4-methoxynaphthalene-1-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only