Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Methoxymethyl)phenylacetic acid, 97%

$94.47 - $325.46

Chemical Identifiers

Chemical Name or Material 4-(Methoxymethyl)phenylacetic acid
CAS 343880-24-0
Molecular Formula C10H12O3
Formula Weight 180.21
MDL Number MFCD08060493
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH50848MD

  SDS
alfa aesar™
H50848MD
250mg Each for $94.47

AAH5084803

  SDS
alfa aesar™
H5084803
1g Each for $325.46
Description & Specifications

Specifications

Formula Weight 180.21
Molecular Formula C10H12O3
MDL Number MFCD08060493
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4-(Methoxymethyl)phenylacetic acid
CAS 343880-24-0
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Quantity 250mg
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.