Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxyindole, 99%

$76.82 - $809.66

Chemical Identifiers

Chemical Name or Material 4-Methoxyindole
CAS 4837-90-5
Molecular Formula C9H9NO
Formula Weight 147.18
MDL Number MFCD00009737
Beilstein 122456
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1906903

  SDS
alfa aesar™
L1906903
1g Each for $212.42

AAL1906906

  SDS
alfa aesar™
L1906906
5g Each for $809.66

AAL19069MD

  SDS
alfa aesar™
L19069MD
250mg Each for $76.82
Description & Specifications

Specifications

Formula Weight 147.18
CAS 4837-90-5
Chemical Name or Material 4-Methoxyindole
Assay 99%
Molecular Formula C9H9NO
MDL Number MFCD00009737
Quantity 1g
Beilstein 122456
Recommended Storage 2° to 8°C
Boiling Point 181° to 183°C (24mmHg)
Melting Point 69° to 71°C
Safety and Handling

2° to 8°C