Please login to your online account to display your discounted pricing

4-Methoxyindole 98%, ACROS Organics™

$198.00 - $198.00

Chemical Identifiers

Chemical Name or Material 4-Methoxyindole
Name Note 98%
CAS 4837-90-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C9H9NO
Formula Weight 147.18
MDL Number MFCD00009737
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC133905000

  SDS
thermo fisher scientific
133905000
Glass bottle 500mg Each for $198.00
Description & Specifications

Specifications

Physical Form Crystalline Powder and/or Chunks
Assay ≥97.5% (GC)
Formula Weight 147.18
Quantity 500mg
Color White
Beilstein 21, II, 44
Melting Point 68° to 72°C
CAS Max % 100.0
Chemical Name or Material 4-Methoxyindole
Infrared Spectrum Authentic
Name Note 98%
Assay Percent Range 98%
Molecular Formula C9H9NO
MDL Number MFCD00009737
Packaging Glass bottle
CAS 4837-90-5
CAS Min % 97.5