missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxycinnamonitrile, cis + trans, 98+%

$91.39 - $327.61

Chemical Identifiers

CAS 28446-68-6
Molecular Formula C10H9NO
Formula Weight 159.19
MDL Number MFCD00001934
Beilstein 2206963
Chemical Name or Material 4-Methoxycinnamonitrile, Cis + Trans
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0077606 SDS Alfa Aesar™
L0077606
5g Each for $91.39
AAL0077614 SDS Alfa Aesar™
L0077614
25g Each for $327.61
Specifications Description & Specifications

Chemical Identifiers

28446-68-6
159.19
2206963
C10H9NO
MFCD00001934
4-Methoxycinnamonitrile, Cis + Trans

Specifications

>110°C (230°F)
≈50° to 55°C
28446-68-6
159.19
2206963
≥98%
130° to 140°C (1mmHg)
5g
C10H9NO
MFCD00001934
4-Methoxycinnamonitrile, Cis + Trans

Safety and Handling

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P321-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Specific treatment (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : GE1459320